Learn More
Invitrogen™ Click-iT™ sDIBO Alkynes for copper-free click chemistry

Experience copper-free 'click chemistry' with Click-IT™ Alexa Fluor™, and sDIBO Alkyne reagents, which are ideal for surface labeling of live cells and minimize damage to enzymes and fluorescent proteins.
Brand: Invitrogen™ C20020
Description
Achieve efficient labeling of azides with Click-It sDIBO Alkynes. Our Click-IT sDIBO alkynes enable the labeling of azido labeled targets without the need for copper catalysis and are available with a variety of Alexa Fluor or biotin labels, as well as amine or thiol reactive groups for creating your own conjugates. Macromolecules that have been azido-modified enzymatically, chemically, or metabolically can now be labeled easily, yielding soluble bioconjugates.
Click-iT sDIBO Alkyne labels react with azides via a copper-free Click chemistry reaction to form a stable triazole linkage with an Alexa Fluor, Biotin, or label of your choice. Copper-free click reactions using our sDIBO alkynes are ideal for surface labeling of live cells; they also minimize damage to fluorescent proteins like GFP or R-PE. For intracellular labeling, the nonspecific binding of DIBO alkynes creates a background signal, and a labeling strategy using picolyl azides is the preferred solution in these instances.
Click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background. Macromolecules that have been azide-modified enzymatically, chemically, or metabolically can be labeled more easily, yielding more soluble bioconjugates with improved biological labeling utility.
To make a concentrated stock solution from 2–5 mM, dissolve the dye-alkyne in high quality anhydrous DMSO. Store frozen, desiccated, and protected from light. Click-iT SDP Ester sDIBO alkyne is susceptible to hydrolysis, so prepare the stock solution fresh on the day of the experiment.

Specifications
Less hazardous | |
Contains 0.5 mg of Alexa Fluor 488 sDIBO Alkyne | |
GFP | |
Blue | |
Dry Ice | |
Alexa Fluor™ 488 | |
Alexa Fluor™, Click-iT™ | |
490 nm |
0.5 mg | |
∼1400 | |
Store at ≤-20°C. Desiccation recommended. Protect material from long-term exposure to light; may be exposed to light for short periods of time | |
Click Chemistry | |
sDIBO Alkyne | |
Green | |
525 nm | |
Azide |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.