Learn More
Tocris Bioscience™ Click N-Acetylmuramic acid - alkyne
Bacterial peptidoglycan derivative; suitable for click-conjugation to fluorescent dyes
Brand:  Tocris Bioscience™ 6798/2
241.70 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18065442
  • 266.00 € / 2mg
Explore more special offers
Description
Description
Derivative of N-acetylmuramic acid (NAM) component of bacterial peptidoglycans. Incorporated into bacterial peptidoglycans during biosynthesis. Suitable for fluorescent labeling of peptidoglycans when click-conjugated to a fluorescent dye.
Specifications
Specifications
(2R)-2-(((3R,4R,5S,6R)-2,5-dihydroxy-6-(hydroxymethyl)-3-(pent-4-ynamido)tetrahydro-2H-pyran-4-yl)oxy)propanoic acid
2 mg
C14H21NO8
2245794-65-2
Glycobiology Probes
95%