missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Clifton™ aluminum Boiling Bath
Regulates from a gentle simmer to a vigorous boil, fitted with concentric ringed lid to vary the hole size. Nickel Electro™ Clifton™ Aluminum Boiling Bath produces steam that provides an effective source of heat for samples that may be damaged by direct flame.
Brand: Nickel Electro™ 1105ER
This item is not returnable.
View return policy
Description
Rectangular, aluminum electrically heated, multi-place baths regulate from a gentle simmer to a vigorous boil, fitted with concentric ringed lid to vary the hole size.The steam produced provides an effective source of heat for samples:
- that may be otherwise thermally damaged by a naked flame.
- used for applications to distill/concentrate liquids, melt agar and other solids.
- metallurgical analysis, protein analysis and bacteriological examinations
- Features an integral constant level device to maintain constant water level during boiling and supplied with a set of rings to vary the diameter of aperture to accommodate different sizes of vessels of evaporating dishes and glassware, including round-bottom flasks.
- Fitted with a sheathed immersion element and a one shot thermal cut out for safety.
- Warranty: 2 year
Specifications
Energy regulator controls the amount of boiling from a gentle simmer to vigorous boil | |
3.12 L | |
Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana | |
1105 ER | |
2 Years | |
400 mm | |
200 mm | |
230 V |
Boiling Bath | |
Simmer to boiling at 100°C | |
Aluminum | |
Rectangular | |
200 x 400 x 90 mm | |
90 mm | |
3.8 kg | |
1000 W |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nickel Electro™ Clifton™ aluminum Boiling Bath > Ambient Temperature Range: Simmer to Boiling at 100°C
Spot an opportunity for improvement?Share a Content Correction