missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Clifton™ aluminum Boiling Bath

Regulates from a gentle simmer to a vigorous boil, fitted with concentric ringed lid to vary the hole size. Nickel Electro™ Clifton™ Aluminum Boiling Bath produces steam that provides an effective source of heat for samples that may be damaged by direct flame.

Brand:  Nickel Electro™ 1105E

 View more versions of this product

Product Code. 12768868

  • 569.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Rectangular, aluminum electrically heated, multi-place baths regulate from a gentle simmer to a vigorous boil, fitted with concentric ringed lid to vary the hole size.

The steam produced provides an effective source of heat for samples:
  • that may be otherwise thermally damaged by a naked flame.
  • used for applications to distill/concentrate liquids, melt agar and other solids.
  • metallurgical analysis, protein analysis and bacteriological examinations
  • Features an integral constant level device to maintain constant water level during boiling and supplied with a set of rings to vary the diameter of aperture to accommodate different sizes of vessels of evaporating dishes and glassware, including round-bottom flasks.
  • Fitted with a sheathed immersion element and a one shot thermal cut out for safety.
  • Warranty: 2 year
Specifications

Specifications

Constant boiling, no control
3.12 L
Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana
1105 E
2 Years
400 mm
200 mm
230 V
Boiling Bath
Constant boiling at 100°C
Aluminum
Rectangular
200 x 400 x 90 mm
90 mm
3.3 kg
1000 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.