Learn More
Nickel Electro™ Clifton™ Bath Lab Armor™
Obtain superior water bath cleaning results when you replace water in your water bath with these thermally efficient pellets that are easy to keep clean. This Clifton™ Bath Armor™ tackles water contamination and protects your water bath from drying out.
Brand:  Nickel Electro™ BA-5
 View more versions of this product
Product Code. 12708898
  • 677.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tackles water contamination by utilizing a dry, metallic thermal media comprised of small beads instead of water
  • Thermally efficient pellets that are easy to keep clean
  • Moisture and gas impermeable
  • High thermal conductivity
  • Smooth and rounded almost fluid like
  • Reduces water consumption
  • Saves the need for racks, floaties and bottle weights
  • Do not autoclave
Specifications
Specifications
4 L
Bath Lab Armor
4 L
Round