Learn More
Nickel Electro™ Clifton™ Digital Thermostirrer with Stainless Steel Tank and Shelf
Get maximum observation of samples, water circulation, and temperature stability with the stirred water bath. Designed for immersion of flasks, bottles, racks in a highly stable temperature environment, this Clifton™ Digital Thermostirrer with Stainless Steel Tank and Shelf includes anti-bacterial technology and other unique features as demanded by many clinical and laboratory environments.
Brand:  Nickel Electro™ NE4-38D
 View more versions of this product
Product Code. 12738908
  • 2105.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stirred Clifton water bath designed for immersion of flasks, bottles, racks in a highly stable temperature environment
  • Thermostirrer is robust - constructed from stainless steel for durability
  • Digital PID temperature control
  • Easy to use controls that wipe clean and have an anti-bacterial finish
  • Sturdy clear co-polyester BPA free tank allows observation of samples
  • Thermostirrer controls and painted surfaces feature “anti-bacterial technology” 
  • When low liquid level is detected, a “FiLL” message automatically appears and heating is suspended
  • Every unit undergoes a factory 2 point calibration procedure 
Specifications
Specifications
Digital Water Bath
CE Approved
Digital LED
NE4-38 D
3 Years
295 x 570 x 200 mm
570 mm
200 mm
295 mm
230 V
50/60 Hz
38 L
5°C to 99°C
Stainless Steel
±0.1°C
330 x 715 x 235 mm
715 mm
235 mm
330 mm
23 kg
1250 W