Learn More
Nickel Electro™ Clifton™ Digital Water Circulator
Get optimum external water circulation and temperature stability with the digital circulator. Clifton™ Digital Water Circulator includes anti-bacterial technology and other unique features as demanded by many clinical and laboratory environments.
Brand:  Nickel Electro™ NE4-P
Product Code. 11766587
  • 1455.00 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
    • Can be fitted to water baths, tanks and containers using accessory Mount Bracket
    • Features stainless steel construction
    • Painted surfaces are powder painted with tough anti-bacterial finish with good chemical resistance
    • Easy to use controls that wipe clean and have an anti-bacterial finish
    • Immersed components are corrosion resistant
    • Low liquid level is detected,  “FiLL” message automatically appears and heating is suspended
    • Features a circulation pump with a capacity 10 litres/minute
Specifications
Specifications
PID Digital
5°C to 99°C
NE4-P
±0.1°C
3 Years
350 mm
230 V
Circulation
Digital LED
±0.1°C
±0.01°C
125 mm
230 V 50/60 Hz
50/60 Hz