Learn More
Clifton™ Digital Shaking Water Bath and Perforated Tray
Get optimum external water circulation and temperature stability with the digital shaker baths.Designed for immersion of flasks, bottles, racks in a highly stable temperature environment; the Clifton™ Digital Shaking Water Bath and Perforated Tray includes anti-bacterial technology and other unique features as demanded by many clinical and laboratory environments.
Brand:  Clifton NE5-10D
Product Code. 11499529
  • 2965.00 € / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Water bath designed for immersion of flasks, bottles, racks in a highly stable temperature environment
  • Precision PID control allows observation of set and actual temperature simultaneously 
  • Automatic tuning option allows bath to learn about your applications, environment and temperatures
  • Painted surfaces feature “anti-bacterial technology
  • Unique, useful stainless steel recessed tray for standing flasks on, prior to, and after immersion
  • High quality corrosion resistant stainless steel tank
  • Bright clean finish housed in a durable paint finished outer case
Specifications
Specifications
10 L
Shaking Water Bath
NE5-10 D
±0.1°C
449 mm
290 mm
300 mm
230 V
50/60 Hz
5°C to 99°C
LED
±0.1°C
3 Years
242 mm
200 mm
230 V, 50/60 Hz, 800 W
800 W
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products