Learn More
Clifton™ RM2 Model Rotary Mixer
Obtain maximum mixing results with the three layered mixing discs ready for tube mixing (sold separately). With the perforated mixing disc it is possible to 'mix and match' a variety of flask clips to suit different applications.
Brand:  Clifton™ RM-2
Product Code. 11796587
  • 510.00 € / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Rotary mixer only. Accessory carousels and a disc with clips are available (sold separately).
Description
Description
  • Small footprint rotator for general use
  • Mixing disc angle is fully adjustable from horizontal to vertical
  • Benchtop or wall mountable.
  • Rotator has option to add accessory heads for mixing micro-tubes, tubes and flasks
  • Heads are quick and easy to change.
  • Accessory carousels and disc with clips are available (sold separately)
  • Suitable for continuous mixing at variable speeds
Specifications
Specifications
5 to 30 rpm
4°C to 60°C
4 kg
3 Years
230 V
Rotary Mixer
For Growth of Mini Preps, Bacterial Culture, Viral Culture, Tissue Culture
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products