missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Stainless Steel Rectangular Electric Boiling Baths

Get optimum boiling results with this rectangular, stainless steel electrically heated boiling bath. Featuring an energy regulator that controls the amount of boiling, this Clifton™ Stainless Steel Rectangular Electric Boiling Bath is ideal for safety.

Brand:  Nickel Electro™ 1102E

 View more versions of this product

Product Code. 12799326

  • 918.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Rectangular, stainless steel electrically heated
  • Multi-place baths with fixed temperature for boiling water
  • Fitted with concentric ringed lids to vary the hole size
  • Features an integral constant level device to maintain constant water level during boiling
  • Fitted with a sheathed immersion element and a one shot thermal cut out for safety
Specifications

Specifications

Constant boiling no control
8.8 L
Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana
1102 E
Rectangular
321 x 425 x 90 mm
90 mm
4.6 kg
Boiling Bath
Constant boiling at 100°C
Stainless Steel
6 x 101 mm Holes
2 Years
425 mm
321 mm
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.