missing translation for 'onlineSavingsMsg'
Learn More

Bryson Packaging™ Cling Film Rolls

Suitable for sealing laboratory containers such as flasks, beakers, bottles, tubes and others. Bryson Packaging™ Cling Film Rolls are supplied in boxes with a cutter edge.

Brand:  Bryson Packaging™ SEL-360-010K

 View more versions of this product

Product Code. 12338039

  • 134.62 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from extruded PVC film, 50μm thick
  • Suitable for use in deep freezers
  • Two-way stretch
  • Staphylococci-proof
  • Heat destruction at 125°C
Specifications

Specifications

Suitable for Sealing laboratory Containers - Flasks, Beakers, Bottles, Tubes and for use in deep freeze
300mm
50 μm
Film
PVC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers