Learn More
Fisherbrand™ Clinical Bundle
GREENER_CHOICE
Optimize your lab efficiency with this comprehensive clinical bundle.
Brand:  Fisherbrand™ 121011F
Product Code. 30130309
  • 1140.00 € / 1 set
Explore more special offers
Description
Description
Spin samples effortlessly with a high-performance centrifuge, keep your tubes organized and accessible with a multi-tube rack, and safeguard your workspace with a durable lab mat. This bundle provides everything you need for streamlined and precise sample handling. This bundle includes the following:
Fisherbrand™ Clinical Centrifuge - #17801727
  • This compact, quiet clinical benchtop centrifuge offers adjustable speeds (500-6,000 RPM), programmable memory, and built-in safety features, maximizing functionality and workspace efficiency.
Fisherbrand™ Clinical 4-Way Tube Rack, Assorted - #16340402
  • This rack holds 13 and 17 mm tubes securely, is made of durable, chemical-resistant polypropylene, and withstands water baths and freezers. Customizable and adaptable for any setup.
Fisherbrand™ Clinical Lab Mat - #16361648
  • This lab mat safely facilitates workflow processes with organization and easy clean up, while keeping benchtops clean and safe from stains or spills.
Specifications
Specifications
Tube Spacers, Adapter Sleeves, 8-Place Rotor, Power Adapter
8 x 3 to 15 mL
6,000 rpm
CE, UKCA, RoHS3, WEEE
93 W
4.5 kg
100/240 V
Clinical Centrifuge
4,668 x g
No
30 sec. to 60 min., or Continuous
9.93 lb.
2°C to 40°C