Learn More
Clinofibrate, MedChemExpress
MedChemExpress Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.
Brand:  Medchem Express HY-13528
Encompass_Preferred
 View more versions of this product
Product Code. 16421897
  • 313.20 € / 50mg
Explore more special offers
Description
Description
Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.
Specifications
Specifications
Clinofibrate
White
C28H36O6
S-8527
CCC(OC1=CC=C(C2(C3=CC=C(OC(CC)(C)C(O)=O)C=C3)CCCCC2)C=C1)(C)C(O)=O
468.58
Research
Metabolism-sugar/lipid metabolism
30299-08-2
Research
50 mg
DMSO : ≥ 30 mg/mL (64.02 mM)
468.58
99.7%
Solid