missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Close-it™ Dispenser
Buerkle™ Close-it™ Dispenser ensures safe storage and simple removal of the labels.
131.00 € - 182.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
10180281
|
Buerkle™
5303-9000 |
95 x 95 mm Close-it Standard Size Labels |
131.00 €
Each |
In Stock
Log in to see stock. |
||||||
10565912
|
Buerkle™
5303-9100 |
150 x 150 mm Close-it Maxi Size Labels |
182.00 €
Each |
In Stock
Log in to see stock. |
||||||
Description
- For close-it standard size 95 x 95mm or close-it Maxi 150 x 150mm
- Ensures safe storage and simple removal of the labels
- Labels can be conveniently written on due to the marking plate
Specifications
Label Dispenser | |
Aluminum |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buerkle™ Close-it™ Dispenser