missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biodesign™ Closure Clamps For Dialysis Tubing
56.05 € - 128.00 €
Specifications
Product Type | Closure Clamp |
---|---|
Quantity | 10 Pk. |
For Use With (Application) | For BioDesign dialysis tubing |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
12777496
|
Biodesign™
D803 |
28.7 mm |
91.70 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12767496
|
Biodesign™
D801 |
21.3 mm |
56.05 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12787496
|
Biodesign™
D805 |
49.5 mm |
128.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Closure Clamp | |
10 Pk. | |
For BioDesign dialysis tubing |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title