missing translation for 'onlineSavingsMsg'
Learn More
Clozapine, 97%, Thermo Scientific™ Product Code.: 15404169

Clozapine, 97%, Thermo Scientific™

Product Code. 15404169
100mg
Click to view available options
Quantity:
25mg
100mg
This item is not returnable. View return policy

Product Code. 15404169

Brand: Thermo Scientific Alfa Aesar J61583.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective muscarinic antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18H19ClN4
Molecular Weight (g/mol) 326.828
MDL Number MFCD00153785
InChI Key ZUXABONWMNSFBN-UHFFFAOYSA-N
Synonym clozapine, leponex, clozapin, clozaril, fazaclo, clorazil, iprox, clozapinum, asaleptin, clozapina
PubChem CID 2818
IUPAC Name 3-chloro-6-(4-methylpiperazin-1-yl)-5H-benzo[b][1,4]benzodiazepine
SMILES CN1CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl

Specifications

Quantity 100mg
UN Number UN2811
Merck Index 14,2423
Formula Weight 326.83
Percent Purity ≥98%
Chemical Name or Material Clozapine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX24K028 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog NumberJ61583.MB,J61583.MC
Lot NumberS27K035 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog NumberJ61583.MB,J61583.MC
Lot NumberQ24K023 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ61583.MB,J61583.MC
Lot NumberP08K035 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog NumberJ61583.MB,J61583.MC
Lot NumberU30J027 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ61583.MB,J61583.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Clozapine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.