missing translation for 'onlineSavingsMsg'
Learn More

Acid Surrogate Standard, SPEX CertiPrep™

Product Code. 12799705
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,6-Tribromophenol118-79-62000μg/mL
2-Fluorophenol367-12-42000μg/mL
Phenol-d613127-88-32000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 118-79-6
Packaging 2mL amber prescored glass ampule
Quantity 1 mL
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C6H3Br3O
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
SMILES OC1=C(Br)C=C(Br)C=C1Br
IUPAC Name 2,4,6-tribromophenol
Molecular Weight (g/mol) 330.80
PubChem CID 1483
ChEBI CHEBI:47696
Methods For US EPA Methods 625, CLP
Analyte or Component Names 2-Fluorophenol; Phenol-d5; 2,4,6-Tribromophenol
Solvent or Matrix Methanol
Chemical Name or Material Acid Surrogate Standard
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • ACID SURRS for CLP 2-88 SOW
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'