missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CMA

Murine-selective STING agonist

204.00 € - 732.00 €

Specifications

Chemical Name or Material 9-Oxo-10(9H)-acridineacetic acid
CAS 38609-97-1
Synonym ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173
Target STING Agonists
Molecular Formula C15H11NO3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18785626
View Documents
Tocris Bioscience™
6677/10
10 mg
204.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18795386
View Documents
Tocris Bioscience™
6677/50
50 mg
732.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • STING agonist
  • Murine-selective (Kd = 3.5 μM)
  • Triggers type I IFN response in murine macrophages, with no effect on human STING signaling
Specifications

Specifications

9-Oxo-10(9H)-acridineacetic acid
ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173
C15H11NO3
98%
38609-97-1
STING Agonists
253.25
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Certificates