missing translation for 'onlineSavingsMsg'
Learn More

Electrothermal™ CMUA Series Stirring/Heating Mantles

Magnetic stirrer and heating mantle in one convenient unit

Brand:  Electrothermal™ 36013-00

779.13 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 15583162

  • 920.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Optimized heat transfer
  • Flexible coiled heating element wraps the heater around round-bottom flask
  • Safe handling is ensured with cool to the touch exterior
Specifications

Specifications

100 mL
17.4625 cm
Flasks
14.9225 cm
230 V
450°C
6.875 in.
67 mm
5.875 in.
17.4625 cm
Stirring Heating Mantle
1 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.