Learn More
Electrothermal™ CMUT Series Multi-Volume Controlled Mantle
Accommodates round bottom flasks between 250 to 1000mL capacity. Electrothermal™ CMUT Series Multi-Volume Contiolled Mantle is used in wet chemistry to heat liquids in round bottom flasks.
Brand:  Electrothermal™ 36013-14
Product Code. 12163781
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Accommodates round bottom flasks between 250 to 1000mL capacity. Electrothermal™ CMUT Series Multi-Volume Contiolled Mantle is used in wet chemistry to heat liquids in round bottom flasks.
  • Metal case is designed to remain “Cool-to-the-touch” when in operation.
  • For added safety, it has a grounded stainless steel earth screen covering the element and is double fused.
  • Multiple volume round bottom flask capacity between 250 to 1000mL
  • Element temperatures up to 450°C
  • Built-in energy regulator
  • Indicator lamps for power and heater operation
  • Replaceable insulated heater cartridge
  • Rugged and easy to clean powder coated aluminum outer casing
  • Non-skid feet and support rod bracket (Support rods not included)
  • Flexible coiled heating elements absorb shock, minimising the risk of flask breakage
  • Heating element is attached to a rigid thermal insulating cartridge to ensure maximum heat transfer and support
Specifications
Specifications
250 to 1000 mL
230 V
1.5kg
50/60 Hz
220 x 220 x 170mm
300W
450°C