missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKSR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | CNKSR3 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892564
|
Proteintech
14084-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882564
|
Proteintech
14084-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.Specifications
CNKSR3 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q5SGD7, Q6P9H4, Q8BMA3 | |
154043, 215748, 308113 | |
CNKSR3 Fusion Protein Ag5213 | |
Primary | |
-20°C | |
CNKSR3 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CNK homolog protein 3, CNK3, CNKSR family member 3, CNKSR3, Connector enhancer of KSR 3, MAGI1, Maguin like protein | |
CNKSR3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title