missing translation for 'onlineSavingsMsg'
Learn More

CNQX, Tocris Bioscience™

Product Code. 18014032
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective non-NMDA iGluR antagonist

CNQX is an AMPA and kainate receptor antagonist (IC50 values are 0.3 μM and 1.5 μM for AMPA and kainate receptors, respectively). CNQX is also an antagonist at the glycine modulatory site on the NMDA receptor complex (IC50 = 25 μM). CNQX can be used to isolate GABAA receptor-mediated spontaneous inhibitory postsynaptic currents and antagonizes non-NMDA receptor-mediated responses in cultured cerebellar granule cells. CNQX shows neuroprotective effects in models of ischemia and inhibits seizure-like activity in hippocampal neurons.

CNQX Disodium Salt (Cat. No. 1045) also available.

Chemical Identifiers

CAS 115066-14-3
Molecular Formula C9H4N4O4
Molecular Weight (g/mol) 232.155
InChI Key RPXVIAFEQBNEAX-UHFFFAOYSA-N
Synonym cnqx, 6-cyano-7-nitroquinoxaline-2,3-dione, unii-6ote87sccw, 7-nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 6ote87sccw, 3h cnqx, 6-cyano-2,3-dihydroxy-7-nitroquinoxaline, 1,4-dihydro-2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile, 6-quinoxalinecarbonitrile, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile
PubChem CID 3721046
ChEBI CHEBI:34468
IUPAC Name 7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile
SMILES C1=C(C(=CC2=C1NC(=O)C(=O)N2)[N+](=O)[O-])C#N

Specifications

Quantity 10 mg
Formula Weight 232.16
Percent Purity >99%
Chemical Name or Material CNQX
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.