Learn More
Eppendorf™ CO22 Bottle Automatic Change-over Controller
For Use With: Innova CO-170, Innova CO-14 , Innova C-48 and Excella ECO-170 Incubators
Brand:  Eppendorf™ P0628-5000
Product Code. 11319944
  • 1751.98 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CO2 Bottle Automatic Changeover Controller
  • Accessory for Innova CO-170, Innova CO-14 , Innova C-48 and Excella ECO-170 Incubators
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Innova CO-170, Innova CO-14 , Innova C-48 and Excella ECO-170 Incubators
Accessory, Cylinder Automatic Change-over Controller
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products