missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Distilled Water for CO2 Incubators, 2 L

Fisherbrand™ water for CO2 Incubators is free of all contaminants and exceeds the specifications set by CO2 Incubator manufacturers.

Brand:  Fisherbrand™ 12-555-881

77.90 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 17856704

  • 82.00 € / 2 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Fisherbrand™ water for CO2 Incubators is double distilled, 0.1um filter sterilized, has a pH of 7-9 with a conductivity of 1-20 microSiemens (Resistivity of 50K-1 M ohm) and is terminal sterilized.

  • Double distilled, 0.1um filter sterilized
  • Has a pH of 7-9 with a conductivity of 1-20 microSiemens (Resistivity of 50K-1 M ohm)
  • Terminal sterilized
Specifications

Specifications

2 L
CO2 Incubators
Sterile Distilled Water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.