Learn More
TFA Dostmann CO2 Monitor AIRCO2NTROL UP Single Beam
The CO2 Monitor AIRCO2NTROL UP Single Beam measures the CO2 concentration in rooms. Its large display shows the CO2 content of your room air, the temperature and humidity, as well as an extended traffic light indication. Additionally, it stores the maximum and minimum values.
Brand:  TFA Dostmann 31.5011.10
Product Code. 17931322
Description
Description
Indoor air quality is becoming increasingly important. After all, a good indoor climate promotes health and well-being. It also improves performance. This is particularly important in schools, offices and public facilities. Basically, anywhere where many people are present at the same time. The higher the concentration, the worse the air.
CO2 remains below the radar of the human senses. We can neither see nor smell it. We can only feel the effects such as tiredness and dizziness. Or those of a viral disease. This is because the likelihood of transmission increases with CO2 exposure: the more exhaled air there is in the room, the greater the risk of becoming infected with the coronavirus via aerosols, for example. With the AIRCO2NTROL UP single beam CO2 monitor, you can keep a constant eye on the air quality. The display shows the current CO2 content of the room air. The temperature and humidity are also shown in the lower display area. With your new air control system, you can now ventilate in good time and constantly maintain a good indoor climate.
How does CO2 monitoring work with the AIRCO2NTROL UP Single Beam from TFA? An NDIR single beam sensor measures the current CO2 content in ppm (parts per million). The exact result is then visualized on the display, as a measured value and as a bar on a circular, graphical display. A traffic light display also visualizes the value. Green indicates good air. Yellow, blue and red warn of increasingly bad air. Values up to 600 ppm are considered ideal. The sensor of this device has a measuring range of 400 ppm to 5000 ppm. An acoustic alarm supports you during monitoring. You can set the upper limit for the alarm signal yourself. Maximum and minimum values for CO2, room temperature and humidity for the last 24 hours are stored and can be called up at the touch of a button.
The result: many valuable measurement results at a glance. So it's good that the CO2 monitor scores with a high-contrast display in smartphone design. Thanks to the integrated stand, the device is stable and can be placed anywhere. A 1.5 m USB cable and power supply unit complete the scope of delivery.
  • For the monitoring of CO2 concentration-levels in rooms
  • Storage of highest and lowest values of CO2, temperature and humidity
  • NDIR Single Beam sensor
  • Large CO2 display with extended traffic light indication (green/yellow/blue/red)
  • Display of indoor temperature (°C/°F) and humidity
  • Alarm function for CO2 concentration
  • High contrast display with 4 brightness levels
  • Manual calibration function for CO2 sensor
  • Power connection via USB Type-C cable and power supply (included) or a suitable USB power source
Specifications
Specifications
Carbon Dioxide
Portable Indoor Air Quality CO2 Meter
5 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TFA Dostmann CO2 Monitor AIRCO2NTROL UP Single Beam > Detectable Analytes: Carbon Dioxide