Learn More
Cobalt aluminum oxide, Co 39-42%, Thermo Scientific Chemicals
43.90 € - 153.00 €
Chemical Identifiers
CAS | 12672-27-4 |
---|---|
Molecular Formula | Al2Co2O5 |
Molecular Weight (g/mol) | 251.824 |
MDL Number | MFCD00016016 |
InChI Key | CRHLEZORXKQUEI-UHFFFAOYSA-N |
Synonym | cobalt aluminate, aluminum cobalt oxide, aluminum cobalt oxide al2coo4, cobalt aluminate 100g, dialuminium cobalt tetraoxide, dialuminum; cobalt 2+ ; oxygen 2-, dialuminium 3+ dicobalt 2+ pentakis oxide 2-, dialuminium 3+ ion bis ? 2-cobalt 2+ ion pentaoxidandiide |
PubChem CID | 159384 |
IUPAC Name | dialuminum;cobalt(2+);oxygen(2-) |
SMILES | [O-2].[O-2].[O-2].[O-2].[O-2].[Al+3].[Al+3].[Co+2].[Co+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363387
|
Thermo Scientific Alfa Aesar
017686.22 |
100 g |
43.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353387
|
Thermo Scientific Alfa Aesar
017686.36 |
500 g |
153.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in the catalytic deposition of nitrous oxide over calcinated cobalt aluminum hydrotalcites. The activity increased with an increase in cobalt concentration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12672-27-4 | |
251.824 | |
CRHLEZORXKQUEI-UHFFFAOYSA-N | |
159384 | |
[O-2].[O-2].[O-2].[O-2].[O-2].[Al+3].[Al+3].[Co+2].[Co+2] |
Al2Co2O5 | |
MFCD00016016 | |
cobalt aluminate, aluminum cobalt oxide, aluminum cobalt oxide al2coo4, cobalt aluminate 100g, dialuminium cobalt tetraoxide, dialuminum; cobalt 2+ ; oxygen 2-, dialuminium 3+ dicobalt 2+ pentakis oxide 2-, dialuminium 3+ ion bis ? 2-cobalt 2+ ion pentaoxidandiide | |
dialuminum;cobalt(2+);oxygen(2-) |
Specifications
12672-27-4 | |
Al2Co2O5 | |
cobalt aluminate, aluminum cobalt oxide, aluminum cobalt oxide al2coo4, cobalt aluminate 100g, dialuminium cobalt tetraoxide, dialuminum; cobalt 2+ ; oxygen 2-, dialuminium 3+ dicobalt 2+ pentakis oxide 2-, dialuminium 3+ ion bis ? 2-cobalt 2+ ion pentaoxidandiide | |
CRHLEZORXKQUEI-UHFFFAOYSA-N | |
dialuminum;cobalt(2+);oxygen(2-) | |
159384 | |
Co 39 to 41% | |
Cobalt aluminum oxide |
Powder | |
MFCD00016016 | |
Insoluble in water. | |
[O-2].[O-2].[O-2].[O-2].[O-2].[Al+3].[Al+3].[Co+2].[Co+2] | |
251.824 | |
176.89 | |
Odorless |
Safety and Handling
GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P272-P280-P281-P285-P302+P352-P304+P341-P305+P351+P338-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H319-H334-H341-H350-H360
missing translation for 'einecsNumber' : 235-762-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Cobalt aluminum oxide
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only