missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Chloride Hexahydrate, >98%, (ACS Reagent Grade), MP Biomedicals

92.60 €

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430592
View Documents
MP Biomedicals
0219403580
100 g
92.60 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7791-13-1
237.92
GFHNAMRJFCEERV-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cl2CoH12O6
MFCD00149652
λ²-cobalt(2+) hexahydrate dichloride

Specifications

7791-13-1
Cl2CoH12O6
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
237.9
Cobalt Chloride Hexahydrate
>98%
MFCD00149652
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
237.92
ACS Reagent
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

Product Identifier
  • Cobalt Chloride Hexahydrate, ACS

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : cobalt chloride hexahydrate

SDS
Documents

Documents

Product Certifications