missing translation for 'onlineSavingsMsg'
Learn More

Cobalt foil, 0.025mm (0.001in), 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11309317
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Quantity 25 x 25 mm
Percent Purity 99.95%
Odor Odorless
Weight ≈0.14g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Cobalt foil, 0.025mm (0.001 in.)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'