missing translation for 'onlineSavingsMsg'
Learn More

Cobalt foil, 0.127mm (0.005in) thick, 50x50mm (2x2in), 99.9% (metals basis), Thermo Scientific Chemicals

172.86 € - 683.40 €

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15496305
View Documents
Thermo Scientific Alfa Aesar
039715.KG
1 Pc.
172.86 €
Each
Please to purchase this item. Need a web account? Register with us today!
15406315
View Documents
Thermo Scientific Alfa Aesar
039715.KJ
5 Pc.
683.40 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-48-4
58.93
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
CHEBI:27638
[Co]

Specifications

7440-48-4
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
104730
99.9%
≈2.83g/50x50mm
(metals basis)
Co
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
[Co]
58.93
CHEBI:27638
Odorless
Foil
Cobalt foil, 0.127mm (0.005 in.) thick, 50x50mm (2x2 in.)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H350

missing translation for 'einecsNumber' : 231-158-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only