missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals

Product Code. 11490817
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

Solubility
Soluble in water, alcohol and dilute acids.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-48-7
Molecular Formula C4H6CoO4
Molecular Weight (g/mol) 177.02
MDL Number MFCD00008689
InChI Key QAHREYKOYSIQPH-UHFFFAOYSA-L
Synonym cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
PubChem CID 6277
ChEBI CHEBI:85138
IUPAC Name cobalt(2+);diacetate
SMILES [Co++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 298°C (decomposition)
Odor Odorless
Linear Formula Co(OOCCH3)2
Quantity 50 g
UN Number UN3077
Sensitivity Hygroscopic
Merck Index 14,2433
Solubility Information Soluble in water,alcohol and dilute acids.
Formula Weight 177.02
Percent Purity ≥98%
Chemical Name or Material Cobalt(II) acetate, anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'