Learn More
Cobalt(II) acetate tetrahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
39.20 € - 312.00 €
Chemical Identifiers
CAS | 6147-53-1 |
---|---|
Molecular Formula | C4H14CoO8 |
Molecular Weight (g/mol) | 249.081 |
MDL Number | MFCD00149569 |
InChI Key | ZBYYWKJVSFHYJL-UHFFFAOYSA-L |
Synonym | cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 |
PubChem CID | 61372 |
IUPAC Name | cobalt(2+);diacetate;tetrahydrate |
SMILES | CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11478470
|
Thermo Scientific Alfa Aesar
044344.14 |
25 g |
39.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468470
|
Thermo Scientific Alfa Aesar
044344.22 |
100 g |
105.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488470
|
Thermo Scientific Alfa Aesar
044344.36 |
500 g |
312.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6147-53-1 | |
249.081 | |
ZBYYWKJVSFHYJL-UHFFFAOYSA-L | |
61372 | |
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] |
C4H14CoO8 | |
MFCD00149569 | |
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 | |
cobalt(2+);diacetate;tetrahydrate |
Specifications
∼140°C (in -4H2O) | |
Red | |
C4H14CoO8 | |
MFCD00149569 | |
3731397 | |
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 | |
ZBYYWKJVSFHYJL-UHFFFAOYSA-L | |
cobalt(2+);diacetate;tetrahydrate | |
61372 | |
98.0 to 102.0% | |
Odorless | |
Hygroscopic | |
Cobalt (II) acetate tetrahydrate |
6147-53-1 | |
Crystalline | |
Co(OOCCH3)2·4H2O | |
UN3077 | |
14,2433 | |
Soluble in water,alcohol,dilute acids and phenyl acetate. | |
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] | |
249.081 | |
249.08 (177.02 Anhydrous) | |
ACS | |
1.542 | |
1.71 g/mL |
Safety and Handling
GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c
H302-H317-H334-H335-H341-H350i-H360F
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 200-755-8
missing translation for 'rtecsNumber' : AG3325000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only