missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide hydrate

Product Code. p-46400 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10297660

Brand: Thermo Scientific Acros 318541000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85017-77-2 | Br2Co | 218.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85017-77-2
Molecular Formula Br2Co
Molecular Weight (g/mol) 218.74
MDL Number MFCD00016017
InChI Key BZRRQSJJPUGBAA-UHFFFAOYSA-L
Synonym cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci
PubChem CID 16212115
IUPAC Name λ²-cobalt(2+) dibromide
SMILES [Co++].[Br-].[Br-]

Specifications

Color Violet
Physical Form Crystals
Quantity 100 g
Linear Formula CoBr2·xH2O
Solubility Information Solubility in water: soluble in water
Formula Weight 218.75
Percent Purity 17.0 to 20.3% (Co)
Packaging Plastic bottle
Chemical Name or Material Cobalt(II) bromide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.