missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide hydrate, Thermo Scientific Chemicals

Product Code. 11309387
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For Hygrometers, Catalysts

Hygrometers, catalystsCobalt(II) bromide hydrate is mainly used as a catalyst in organic synthesis, like C-metal, C-heteroatom and C-C bond formations, addition to unsaturated bonds, cyclizations, polymerization, oxidations reductions and carbonylations. It reacts with ammonia to form coordination compound bromopentaamminecobalt(III) bromide. It reacts with triphenylphosphine and form complexes of cobalt(II) bromide, which used as a catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85017-77-2
Molecular Formula Br2Co
Molecular Weight (g/mol) 218.74
MDL Number MFCD00016017
InChI Key BZRRQSJJPUGBAA-UHFFFAOYSA-L
Synonym cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci
PubChem CID 16212115
SMILES [Co++].[Br-].[Br-]

Specifications

Melting Point 47°C to 48°C (decomposition)
Color Purple-Red
Physical Form Crystalline Granules
Quantity 250 g
Solubility Information Soluble in water.
IUPAC Name dibromocobalt;hydrate
Formula Weight 218.75 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Red-purple crystalline granules
Density 2.46
Chemical Name or Material Cobalt(II) bromide hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.