missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide hydrate, Thermo Scientific Chemicals

For Hygrometers, Catalysts

87.94 € - 321.60 €

Chemical Identifiers

CAS 85017-77-2
Molecular Formula Br2Co
Molecular Weight (g/mol) 218.74
MDL Number MFCD00016017
InChI Key BZRRQSJJPUGBAA-UHFFFAOYSA-L
Synonym cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci
PubChem CID 16212115
SMILES [Co++].[Br-].[Br-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11399377
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
040225.18
50 g
87.94 €
50g
Please to purchase this item. Need a web account? Register with us today!
11309387
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
040225.30
250 g
321.60 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hygrometers, catalystsCobalt(II) bromide hydrate is mainly used as a catalyst in organic synthesis, like C-metal, C-heteroatom and C-C bond formations, addition to unsaturated bonds, cyclizations, polymerization, oxidations reductions and carbonylations. It reacts with ammonia to form coordination compound bromopentaamminecobalt(III) bromide. It reacts with triphenylphosphine and form complexes of cobalt(II) bromide, which used as a catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

85017-77-2
218.74
BZRRQSJJPUGBAA-UHFFFAOYSA-L
16212115
Br2Co
MFCD00016017
cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci
[Co++].[Br-].[Br-]

Specifications

47°C to 48°C (decomposition)
Purple-Red
Br2Co
cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci
BZRRQSJJPUGBAA-UHFFFAOYSA-L
λ²-cobalt(2+) dibromide
16212115
Odorless
Red-purple crystalline granules
Cobalt(II) bromide hydrate
85017-77-2
Crystalline Granules
MFCD00016017
Soluble in water.
[Co++].[Br-].[Br-]
218.74
218.75 (Anhydrous)
Hygroscopic
2.46 g/cm3
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 232-166-7

missing translation for 'rtecsNumber' : GF9595000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.