missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

134.00 € - 465.00 €

Chemical Identifiers

CAS 7789-43-7
Molecular Formula Br2Co
Molecular Weight (g/mol) 218.741
MDL Number MFCD00016017
InChI Key BZRRQSJJPUGBAA-UHFFFAOYSA-L
Synonym cobalt bromide, cobalt ii bromide, cobalt bromide cobr2, cobalt dibromide, cobaltous bromide, hsdb 998, cobalt bromide ous
PubChem CID 24610
IUPAC Name dibromocobalt
SMILES [Co](Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313888
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040319.03
1 g
134.00 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11303888
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040319.06
5 g
465.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used primarily as a catalyst in some processes. Chiefly used in hygrometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-43-7
218.741
BZRRQSJJPUGBAA-UHFFFAOYSA-L
24610
[Co](Br)Br
Br2Co
MFCD00016017
cobalt bromide, cobalt ii bromide, cobalt bromide cobr2, cobalt dibromide, cobaltous bromide, hsdb 998, cobalt bromide ous
dibromocobalt

Specifications

678°C (in N2)
Green
99.99% (Metal basis)
MFCD00016017
Soluble in water. Soluble in methyl acetate,ether,alcohol,acetone.
[Co](Br)Br
218.741
218.75
Green (powder/flake/crystalline/beads, etc.)
Cobalt(II) bromide
7789-43-7
Beads
Br2Co
cobalt bromide, cobalt ii bromide, cobalt bromide cobr2, cobalt dibromide, cobaltous bromide, hsdb 998, cobalt bromide ous
BZRRQSJJPUGBAA-UHFFFAOYSA-L
dibromocobalt
24610
Hygroscopic
4.909 g/cm3
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P272-P280-P281-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c

H302-H315-H317-H319-H334-H350

missing translation for 'einecsNumber' : 232-166-7

missing translation for 'rtecsNumber' : GF9595000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only