missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride, 0.5M aq. soln., Thermo Scientific™

34.00 € - 52.30 €

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15495509
View Documents
Thermo Scientific Alfa Aesar
J63246.AD
50 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15405519
View Documents
Thermo Scientific Alfa Aesar
J63246.AE
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7791-13-1
237.92
GFHNAMRJFCEERV-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cl2CoH12O6
MFCD00149652
λ²-cobalt(2+) hexahydrate dichloride

Specifications

7791-13-1
Liquid
CoCl2
UN3082
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
237.92
0.5M aq. soln.
Colorless
Cl2CoH12O6
MFCD00149652
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
129.84
Cobalt(II) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications