missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent, Thermo Scientific Chemicals

50.15 € - 316.58 €

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10454321
View Documents
Thermo Scientific Acros
423570050
5 g Glass bottle
50.15 €
5g
Please to purchase this item. Need a web account? Register with us today!
10414711
View Documents
Thermo Scientific Acros
423571000
100 g Plastic Bottle
87.64 €
100g
Please to purchase this item. Need a web account? Register with us today!
10361691
View Documents
Thermo Scientific Acros
423575000
500 g Plastic Bottle
316.58 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

7791-13-1
237.92
GFHNAMRJFCEERV-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cl2CoH12O6
MFCD00149652
λ²-cobalt(2+) hexahydrate dichloride

Specifications

86.0°C
Lilac
98.0 to 102% (Complexometry)
CoCl2·6H2O
01,155; 06,127; 11,479
Solubility in water: 970g/L (20°C)
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
237.92
98 to 102%
Negligible
7791-13-1
Fine Crystalline Powder
Cl2CoH12O6
MFCD00149652
15, 2421
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
237.93
ACS Reagent
Cobalt(II) chloride hexahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing genetic defects.
May cause cancer by inhalation.
May cause an allergic skin reaction.
May cause allergy or ast

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:

GHS Signal Word: Danger

missing translation for 'rtecsNumber' : GG0200000

Product Identifier
  • Cobalt(II) chloride hexahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only