missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Sensitivity: Hygroscopic; m.p. 52-56°C -4H2O, 100°C -5H2O, 120-140°C -6H2O

109.55 € - 348.74 €

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311119
View Documents
Thermo Scientific Alfa Aesar
011344.30
250 g
109.55 €
250g
Please to purchase this item. Need a web account? Register with us today!
11301119
View Documents
Thermo Scientific Alfa Aesar
011344.A1
1 kg
348.74 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cobalt(II) chloride hexahydrate is used as a humidity indicator in weather instruments. It finds use in electroplating of cobalt, in invisible ink, in organic chemistry and is a precursor to cobaltocene(bis (cyclopentadienyl) cobalt(II)).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7791-13-1
237.92
GFHNAMRJFCEERV-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cl2CoH12O6
MFCD00149652
λ²-cobalt(2+) hexahydrate dichloride

Specifications

87°C
Purple-Red
99.9% (Metals basis)
MFCD00149652
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
237.93 (129.84 Anhydrous)
Hygroscopic
Cobalt(II) chloride hexahydrate
7791-13-1
Crystals
Cl2CoH12O6
Soluble in water,alcohol,acetone,ether and glycerol.
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
237.92
Odorless
1.924
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302+H332-H317-H334-H341-H350i-H360F

missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III

missing translation for 'einecsNumber' : 231-589-4

missing translation for 'rtecsNumber' : GG0200000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.