missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Chloride Hexahydrate, ACS Reagent, Honeywell Fluka™

Product Code. 15622760
Click to view available options
Quantity:
5 g
100 g
500 g

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Quantity 100 g
Linear Formula CoCl2 · 6H2O
UN Number 3077
Formula Weight 237.93g/mol
Percent Purity 98%
Grade ACS Reagent
Packaging Plastic Bottle
Vapor Pressure 40mmHg (0°C)
Chemical Name or Material Cobalt(Ii) Chloride Hexahydrate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : cobalt dichloride hexahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'