missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

94.30 € - 336.00 €

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11330949
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010692.09
10 g
94.30 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11320949
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010692.18
50 g
336.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cobalt(II) chloride hexahydrate has been used to study apoptotic effects in HepG2 cells. Also reported as a stabilizer for reconstitution of the radiopharmaceutical technetium-99m exametazime. It has been used in studies to examine heat shock protein expression of inflamed skin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7791-13-1
237.92
GFHNAMRJFCEERV-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cl2CoH12O6
MFCD00149652
λ²-cobalt(2+) hexahydrate dichloride

Specifications

87°C
Purple-Red
Crystalline Aggregates
CoCl2·6H2O
UN3077
Soluble in water,acetone,ether,and alcohols.
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
237.92
99.998%
Odorless
1.924 g/mL
Cobalt(II) chloride hexahydrate
7791-13-1
4.9
Cl2CoH12O6
MFCD00149652
14,2437
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
237.93 (129.84 Anhydrous)
Puratronic™
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302+H332-H317-H334-H341-H350i-H360F

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 231-589-4

missing translation for 'rtecsNumber' : GG0200000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only