missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Cobalt(II) chloride in vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1049°C |
Melting Point | 735°C |
Color | Blue |
Physical Form | Beads |
Quantity | 1 g |
Linear Formula | CoCl2 |
UN Number | UN3077 |
Merck Index | 14,2437 |
Formula Weight | 129.84 |
Percent Purity | 99.9% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'