Learn More
Cobalt(II) chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals
Sensitivity: Hygroscopic; 3.367. Decomposes at 400°C on long heating in air
69.45 € - 679.38 €
Chemical Identifiers
CAS | 7646-79-9 |
---|---|
Molecular Formula | Cl2Co |
Molecular Weight (g/mol) | 129.83 |
MDL Number | MFCD00010938 |
InChI Key | GVPFVAHMJGGAJG-UHFFFAOYSA-L |
Synonym | cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride |
IUPAC Name | λ²-cobalt(2+) dichloride |
SMILES | [Cl-].[Cl-].[Co++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366928
|
Thermo Scientific Alfa Aesar
013666.03 |
1 g |
69.45 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356928
|
Thermo Scientific Alfa Aesar
013666.06 |
5 g |
198.99 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346928
|
Thermo Scientific Alfa Aesar
013666.14 |
25 g |
679.38 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) chloride in vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7646-79-9 | |
129.83 | |
GVPFVAHMJGGAJG-UHFFFAOYSA-L | |
λ²-cobalt(2+) dichloride |
Cl2Co | |
MFCD00010938 | |
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride | |
[Cl-].[Cl-].[Co++] |
Specifications
1049°C | |
7646-79-9 | |
Beads | |
CoCl2 | |
UN3077 | |
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride | |
[Cl-].[Cl-].[Co++] | |
129.83 | |
99.9% | |
Ampouled under Argon | |
3.367 | |
Cobalt(II) chloride, ultra dry |
735°C | |
Blue | |
Cl2Co | |
MFCD00010938 | |
14,2437 | |
GVPFVAHMJGGAJG-UHFFFAOYSA-L | |
λ²-cobalt(2+) dichloride | |
129.84 | |
Odorless | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H334-H350-H360-H341-H302-H317;
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'rtecsNumber' : GF9800000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.