missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) citrate dihydrate, 98%, Thermo Scientific Chemicals

38.19 € - 102.51 €

Chemical Identifiers

CAS 6424-15-3
Molecular Formula C12H20Co3O16
Molecular Weight (g/mol) 597.076
MDL Number MFCD02684007
InChI Key SKMIIAIULKZNRQ-UHFFFAOYSA-N
Synonym cobalt ii citrate dihydrate
PubChem CID 131882668
IUPAC Name cobalt;2-hydroxypropane-1,2,3-tricarboxylic acid;dihydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.[Co].[Co].[Co]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11481087
View Documents
Thermo Scientific Alfa Aesar
B24088.14
25 g
38.19 €
25g
Please to purchase this item. Need a web account? Register with us today!
11491087
View Documents
Thermo Scientific Alfa Aesar
B24088.22
100 g
102.51 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6424-15-3
597.076
SKMIIAIULKZNRQ-UHFFFAOYSA-N
131882668
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.[Co].[Co].[Co]
C12H20Co3O16
MFCD02684007
cobalt ii citrate dihydrate
cobalt;2-hydroxypropane-1,2,3-tricarboxylic acid;dihydrate

Specifications

∼150°C (decomposition)
C12H20Co3O16
UN3077
SKMIIAIULKZNRQ-UHFFFAOYSA-N
cobalt;2-hydroxypropane-1,2,3-tricarboxylic acid;dihydrate
131882668
98%
6424-15-3
MFCD02684007
cobalt ii citrate dihydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.[Co].[Co].[Co]
597.076
591.02 (554.99 Anhydrous)
Cobalt(II) citrate dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H341-H350-H360

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 212-751-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only