missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) fluoride, 99%, Thermo Scientific Chemicals

Product Code. 10562091
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-17-2
Molecular Formula CoF2
Molecular Weight (g/mol) 96.96
MDL Number MFCD00010941
InChI Key YCYBZKSMUPTWEE-UHFFFAOYSA-L
Synonym cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry
PubChem CID 24820
IUPAC Name difluorocobalt
SMILES F[Co]F

Specifications

Boiling Point 1400°C
Melting Point 1200°C
Color Pink
Physical Form Powder
Quantity 25 g
Assay Percent Range 98.5% min. (ex F)
Linear Formula CoF2
Merck Index 15, 2424
Formula Weight 96.96
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Cobalt(II) fluoride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cobalt(II) fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.