Learn More
Cobalt(II) fluoride, anhydrous, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 10026-17-2 |
---|---|
Molecular Formula | CoF2 |
Molecular Weight (g/mol) | 96.93 |
MDL Number | MFCD00010941 |
InChI Key | YCYBZKSMUPTWEE-UHFFFAOYSA-L |
Synonym | cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry |
PubChem CID | 24820 |
IUPAC Name | difluorocobalt |
SMILES | F[Co]F |
Description
Cobalt(II) fluoride can be used as a catalyst to alloy metals. It is also used for optical deposition, of which it tremendously improves optical quality. The compound may be used in dental care.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10026-17-2 | |
96.93 | |
YCYBZKSMUPTWEE-UHFFFAOYSA-L | |
24820 | |
F[Co]F |
CoF2 | |
MFCD00010941 | |
cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry | |
difluorocobalt |
Specifications
1400°C | |
10026-17-2 | |
CoF2 | |
UN2923 | |
cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry | |
YCYBZKSMUPTWEE-UHFFFAOYSA-L | |
difluorocobalt | |
24820 | |
98% | |
Hygroscopic | |
Cobalt(II) fluoride, Anhydrous |
1200°C | |
Powder | |
MFCD00010941 | |
14,2440 | |
Soluble in water and hydrofluoric acid. insoluble in alcohol,ether,benzene. | |
F[Co]F | |
96.93 | |
96.93 | |
Odorless | |
4.46 g/mL |
Safety and Handling
GHS H Statement
H301-H314-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 233-061-9
missing translation for 'rtecsNumber' : GG0770000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only