Learn More
Cobalt(II) fluoride tetrahydrate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
58.45 € - 170.00 €
Chemical Identifiers
CAS | 13817-37-3 |
---|---|
Molecular Formula | CoF2H8O4 |
Molecular Weight (g/mol) | 168.99 |
MDL Number | MFCD00149656 |
InChI Key | MNHUTFNEBYCZAF-UHFFFAOYSA-L |
Synonym | cobalt 2+ tetrahydrate difluoride, acmc-1btz4, cobaltous fluoride 4h2o, cobalt ii ; fluoride tetrahydrate, cobalt 2+ fluoride-water 1/2/4, cobalt fluoride cof2 ,tetrahydrate 8ci,9ci |
PubChem CID | 24884271 |
SMILES | O.O.O.O.[F-].[F-].[Co++] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13817-37-3 | |
168.99 | |
MNHUTFNEBYCZAF-UHFFFAOYSA-L | |
24884271 |
CoF2H8O4 | |
MFCD00149656 | |
cobalt 2+ tetrahydrate difluoride, acmc-1btz4, cobaltous fluoride 4h2o, cobalt ii ; fluoride tetrahydrate, cobalt 2+ fluoride-water 1/2/4, cobalt fluoride cof2 ,tetrahydrate 8ci,9ci | |
O.O.O.O.[F-].[F-].[Co++] |
Specifications
200°C (decomposition) | |
Pink | |
CoF2H8O4 | |
UN2923 | |
cobalt 2+ tetrahydrate difluoride, acmc-1btz4, cobaltous fluoride 4h2o, cobalt ii ; fluoride tetrahydrate, cobalt 2+ fluoride-water 1/2/4, cobalt fluoride cof2 ,tetrahydrate 8ci,9ci | |
O.O.O.O.[F-].[F-].[Co++] | |
168.99 | |
168.99 (96.93 Anhydrous) | |
Puratronic™ | |
Moisture sensitive | |
(metals basis) |
13817-37-3 | |
Powder | |
MFCD00149656 | |
14,2440 | |
MNHUTFNEBYCZAF-UHFFFAOYSA-L | |
cobalt(2+);difluoride;tetrahydrate | |
24884271 | |
99.99% | |
Odorless | |
2.19 g/mL | |
Cobalt(II) fluoride tetrahydrate |
Safety and Handling
GHS H Statement
H301-H314-H318-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P281-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H334-H335-H350
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 233-061-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.