missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) hexafluoro-2,4-pentanedionate hydrate, 97%, Thermo Scientific™

58.00 € - 200.00 €

Chemical Identifiers

CAS 206986-92-7
Molecular Formula C10H6CoF12O5
Molecular Weight (g/mol) 493.07
MDL Number MFCD00150649,MFCD00042510
InChI Key POHWVOSIFCILEF-UHFFFAOYSA-N
Synonym bis hexafluoroacetylacetonato cobalt ii hydrate, cobalt 2+ bis 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, cobalt 2+ ; z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
PubChem CID 16212172
IUPAC Name cobalt;(Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate
SMILES O.[Co].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398718
View Documents
Thermo Scientific Alfa Aesar
041594.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11308728
View Documents
Thermo Scientific Alfa Aesar
041594.06
5 g
200.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

206986-92-7
493.07
POHWVOSIFCILEF-UHFFFAOYSA-N
16212172
O.[Co].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
C10H6CoF12O5
MFCD00150649,MFCD00042510
bis hexafluoroacetylacetonato cobalt ii hydrate, cobalt 2+ bis 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, cobalt 2+ ; z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
cobalt;(Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate

Specifications

173°C to 179°C
Orange
C10H6CoF12O5
bis hexafluoroacetylacetonato cobalt ii hydrate, cobalt 2+ bis 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, cobalt 2+ ; z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
POHWVOSIFCILEF-UHFFFAOYSA-N
cobalt;(Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate
16212172
97%
Cobalt(II) hexafluoro-2,4-pentanedionate hydrate
206986-92-7
Powder
MFCD00150649,MFCD00042510
It is insoluble in water.
O.[Co].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
493.07
473.04 (Anhydrous)
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H315-H319-H317-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c

H315-H317-H319-H335-H350

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.