missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Product Code. 15432457
Click to view available options
Quantity:
50 g
250 g
1000 g

Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-06-1
Molecular Formula Co3O4
Molecular Weight (g/mol) 240.80
MDL Number MFCD00010939
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
PubChem CID 11651651
IUPAC Name oxocobalt;oxo(oxocobaltiooxy)cobalt
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Specifications

Melting Point 900°C (decomposition)
Color Black
Physical Form Powder
Quantity 1000 g
Assay Percent Range 99.7% (Metal basis)
Solubility Information Soluble in acids and alkalies. Insoluble in water.
Formula Weight 240.8
Odor Odorless
Appearance Black powder
Density 6.11
Chemical Name or Material Cobalt(II, III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'