missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals Product Code.: 15442457

Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Product Code. 15442457
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 15442457

Brand: Thermo Scientific Alfa Aesar 045806.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-06-1
Molecular Formula Co3O4
Molecular Weight (g/mol) 240.80
MDL Number MFCD00010939
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
PubChem CID 11651651
IUPAC Name oxocobalt;oxo(oxocobaltiooxy)cobalt
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Specifications

Melting Point 900°C (decomposition)
Color Black
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.7% (Metal basis)
Solubility Information Soluble in acids and alkalies. Insoluble in water.
Formula Weight 240.8
Odor Odorless
Appearance Black powder
Density 6.11 g/cm3
Chemical Name or Material Cobalt(II, III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN24L029 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog Number045806.18,045806.30,045806.0B
Lot NumberZ06J005 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number045806.18,045806.30,045806.0B
Lot NumberS28I002 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog Number045806.18,045806.30,045806.0B
Lot NumberN21F039 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number045806.18,045806.30,045806.0B
Lot NumberW28C069 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045806.18,045806.30,045806.0B

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt