Learn More
Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals
106.53 € - 1087.41 €
Chemical Identifiers
CAS | 1308-06-1 |
---|---|
Molecular Formula | Co3O4 |
Molecular Weight (g/mol) | 240.80 |
MDL Number | MFCD00010939 |
InChI Key | UBEWDCMIDFGDOO-UHFFFAOYSA-N |
Synonym | oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt |
PubChem CID | 11651651 |
IUPAC Name | oxocobalt;oxo(oxocobaltiooxy)cobalt |
SMILES | [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442457
|
Thermo Scientific Alfa Aesar
045806.18 |
50 g |
106.53 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452457
|
Thermo Scientific Alfa Aesar
045806.30 |
250 g |
392.96 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15432457
|
Thermo Scientific Alfa Aesar
045806.0B |
1000 g |
1087.41 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
1308-06-1 | |
240.80 | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
11651651 | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Co3O4 | |
MFCD00010939 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt | |
oxocobalt;oxo(oxocobaltiooxy)cobalt |
Specifications
900°C (decomposition) | |
Black | |
99.7% (Metal basis) | |
MFCD00010939 | |
Soluble in acids and alkalies. Insoluble in water. | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] | |
240.80 | |
240.8 | |
Black powder | |
Cobalt(II, III) oxide |
1308-06-1 | |
Powder | |
Co3O4 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
oxocobalt;oxo(oxocobaltiooxy)cobalt | |
11651651 | |
Odorless | |
6.11 g/cm3 |
Safety and Handling
P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
H334-H350i
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 215-157-2
missing translation for 'rtecsNumber' : GG2500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.